Learn More
Apexbio Technology LLC Pioglitazone HCl, 500mg Cas# 112529-15-4
SDP
Supplier:  Apexbio Technology LLC A4324500
Encompass_Preferred
Pioglitazone HCl is a potent and highly selective agonist of PPARY with IC50 value of 3uM. 112529-15-4. MFCD04975446 Pioglitazone HCl is a potent and highly selective agonist of PPAR with IC50 value of 3M[1].Pioglitazone HCl can increase both insulin-stimulated glucose uptake in peripheral tissues and insulin sensitivity in hepatic and adipose tissue. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8588