Learn More
Medchemexpress LLC Pioglitazone hydrochloride | 112529-15-4 | 100.0% | C19H21ClN2O3S | 5 G
SDP
Catalog No. 5000353319
Encompass_Preferred
Pioglitazone hydrochloride is a potent and selective PPARγ agonist, commonly used in research for ferroptosis, metabolic disease, and cancer. It impacts cellular processes through various pathways including cell cycle/DNA damage, metabolic enzyme/protease, vitamin D related/nuclear receptor, and apoptosis.
  • Functions as a potent and selective PPARγ agonist.
  • Known by synonyms U 72107A and AD 4833.
  • Explored in research areas such as PPAR, ferroptosis, metabolic disease, and cancer.
  • Affects pathways including cell cycle/DNA damage, metabolic enzyme/protease, vitamin D related/nuclear receptor, and apoptosis.
  • Demonstrates activity with human PPARδ and PPARα.
  • Cited in 42 publications.
Catalog No. 50-003-53319 Supplier Medchemexpress LLC Supplier No. HY146015G
Pioglitazone hydrochloride is a potent and selective PPARγ agonist, commonly used in research for ferroptosis, metabolic disease, and cancer. It impacts cellular processes through various pathways including cell cycle/DNA damage, metabolic enzyme/protease, vitamin D related/nuclear receptor, and apoptosis.
  • Functions as a potent and selective PPARγ agonist.
  • Known by synonyms U 72107A and AD 4833.
  • Explored in research areas such as PPAR, ferroptosis, metabolic disease, and cancer.
  • Affects pathways including cell cycle/DNA damage, metabolic enzyme/protease, vitamin D related/nuclear receptor, and apoptosis.
  • Demonstrates activity with human PPARδ and PPARα.
  • Cited in 42 publications.