Learn More
Medchemexpress LLC Pioglitazone hydrochloride | 112529-15-4 | 99.97% | C19H21ClN2O3S | 1 G
SDP
Catalog No. 5000353315
Encompass_Preferred
Pioglitazone hydrochloride is a potent and selective PPARγ agonist with EC50s of 0.93 and 0.99 μM for human and mouse PPARγ, respectively. It targets PPAR and Ferroptosis, and is relevant for research in metabolic disease and cancer.
  • Potent and selective PPARγ agonist
  • Targets PPAR and ferroptosis
  • Research applications in metabolic disease and cancer
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 2 years or -20°C for 1 year (sealed, away from moisture)
Catalog No. 50-003-53315 Supplier Medchemexpress LLC Supplier No. HY146011G
Pioglitazone hydrochloride is a potent and selective PPARγ agonist with EC50s of 0.93 and 0.99 μM for human and mouse PPARγ, respectively. It targets PPAR and Ferroptosis, and is relevant for research in metabolic disease and cancer.
  • Potent and selective PPARγ agonist
  • Targets PPAR and ferroptosis
  • Research applications in metabolic disease and cancer
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 2 years or -20°C for 1 year (sealed, away from moisture)