Learn More
Cayman Chemical Pioglitazone (potassium salt), Purity: ≥98%, CAS: 1266523-09-4, 5 mg
SDP
Catalog No. 502890548
Encompass_Preferred
A PPARγ agonist (EC50 = ~500-600 nM for both human and murine PPARγ); selective for PPARγ over PPARα exhibiting low level activation of PPARα at 1 µM and 5.4-fold activation at a concentration of 10 µM; reduces hyperglycemia, hyperlipidemia, and hyperinsulinemia in a dose-dependent manner in male Wistar fatty rats from 0.3-3 mg/kg; reduces the number of lesions in a TRAP model; decreases production of neuroinflammatory cytokines and reduces immobility in the forced swim and tail suspension tests in a mouse model of chronic mild stress at 2.5 mg/kg
Catalog No. 50-289-0548 Supplier Cayman Chemical Supplier No. 100285 MG
A PPARγ agonist (EC50 = ~500-600 nM for both human and murine PPARγ); selective for PPARγ over PPARα exhibiting low level activation of PPARα at 1 µM and 5.4-fold activation at a concentration of 10 µM; reduces hyperglycemia, hyperlipidemia, and hyperinsulinemia in a dose-dependent manner in male Wistar fatty rats from 0.3-3 mg/kg; reduces the number of lesions in a TRAP model; decreases production of neuroinflammatory cytokines and reduces immobility in the forced swim and tail suspension tests in a mouse model of chronic mild stress at 2.5 mg/kg