Learn More
OHAUS Pioneer PX Precision Balance
Catalog No. 01920220 Shop All OHAUS Products
Change view
Click to view available options
OHAUS Precision Balance
Catalog No.
01-920-220 OHAUS Precision Balance
1 options
Catalog No. 01-920-220 Supplier OHAUS Supplier No. 30699095
Add to Cart
Add to Cart
The Pioneer™ PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings. The PX is affordably priced, and intuitively designed for intelligent operation with a second line display for additional information.
The PX offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and education settings at an economical price point.
  • Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX is durably constructed for versatile, long-term use.
  • Pioneer features a second line display for additional information or guidance, a static removal bar for convenient grounding, and USB connectivity.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 6200 g
Display Type LCD
Readability 0.05 g
Type Balance
Legal for Trade No