Learn More
Medchemexpress LLC PIP-199 | 622795-76-0 | 98.0% | 456.54 | C27H28N4O3 | 1MG
SDP
Catalog No. 5000238579
Encompass_Preferred
PIP-199 is a small-molecule research inhibitor of the RMI-MM2 protein-protein interaction (reported IC50 36 μM). It is intended for in vitro studies of genome instability and to investigate sensitization of tumor cells to DNA crosslinking chemotherapeutics. Supplied as a high-purity sample with supporting safety and quality documents.
Catalog No. 50-002-38579 Supplier Medchemexpress LLC Supplier No. HY1243251MG
PIP-199 is a small-molecule research inhibitor of the RMI-MM2 protein-protein interaction (reported IC50 36 μM). It is intended for in vitro studies of genome instability and to investigate sensitization of tumor cells to DNA crosslinking chemotherapeutics. Supplied as a high-purity sample with supporting safety and quality documents.