Learn More
MSC PIP Arm Sleeve, 18 in, ACP/Kevlar, Cut A5, 1 ea
Catalog No. 5000793305
Encompass
The Kut Gard Single-Ply ACP / Kevlar Blended Sleeve with Cotton Liner is designed for exceptional comfort and superior protection. Its proprietary blend offers twice the cut resistance of traditional Para-Aramid sleeves and is heat resistant up to 350°F, protecting arms from minor heat and burn injuries.
  • Twice the cut resistance compared with traditional Para-Aramid sleeves
  • Heat resistant up to 350°F
  • Exceptional comfort, will not cause skin irritation
  • Cotton lining for superior comfort
  • Washable and dry-cleanable
  • Cut level A5
  • ANSI contact heat level 3
  • Reusable and launderable
  • TAA compliant
Catalog No. 50-007-93305 Supplier MSC Supplier No. 47234638
The Kut Gard Single-Ply ACP / Kevlar Blended Sleeve with Cotton Liner is designed for exceptional comfort and superior protection. Its proprietary blend offers twice the cut resistance of traditional Para-Aramid sleeves and is heat resistant up to 350°F, protecting arms from minor heat and burn injuries.
  • Twice the cut resistance compared with traditional Para-Aramid sleeves
  • Heat resistant up to 350°F
  • Exceptional comfort, will not cause skin irritation
  • Cotton lining for superior comfort
  • Washable and dry-cleanable
  • Cut level A5
  • ANSI contact heat level 3
  • Reusable and launderable
  • TAA compliant