Learn More
Echelon Biosciences Research Labs PIP ARRAY 2 pack
SDP
Catalog No. 50720660
Encompass
PIP Arrays are 4 x 5 cm hydrophobic membranes that have been spotted with a concentration gradient of all eight phosphoinositides. They are used to determine the relative degree of binding of your protein of interest towards one or more of these lipids in a simple protein lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-660 Supplier Echelon Biosciences Research Labs Supplier No. P61002PK
PIP Arrays are 4 x 5 cm hydrophobic membranes that have been spotted with a concentration gradient of all eight phosphoinositides. They are used to determine the relative degree of binding of your protein of interest towards one or more of these lipids in a simple protein lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable