Learn More
MSC PIP Chemical Resistant Gloves, Neoprene/Latex, 3X-Large, 28 mil, 1 Pair
Catalog No. 5000678001
Encompass
These chemical resistant gloves offer reliable protection for tasks requiring dexterity and involving light-to-medium chemical applications.
  • Material: Neoprene over latex
  • Glove size: 3X-Large (numeric size 12)
  • Finish: Raised diamond grip
  • Lining: Flocked
  • Color: Blue
  • Thickness: 28 mil
  • Supported or unsupported: Unsupported
  • Cuff style: Straight
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA cut resistance level 1
Catalog No. 50-006-78001 Supplier MSC Supplier No. 38341939
These chemical resistant gloves offer reliable protection for tasks requiring dexterity and involving light-to-medium chemical applications.
  • Material: Neoprene over latex
  • Glove size: 3X-Large (numeric size 12)
  • Finish: Raised diamond grip
  • Lining: Flocked
  • Color: Blue
  • Thickness: 28 mil
  • Supported or unsupported: Unsupported
  • Cuff style: Straight
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA cut resistance level 1