Learn More
MSC PIP Cleanroom Nitrile Glove, Class 100, Finger Textured, 100/Pk.
Catalog No. 5000792250
Encompass
These nitrile gloves are designed for use in controlled environments such as biotechnology and computer assembly, exceeding standards for Class 100 cleanrooms. They offer excellent grip and tactile sensitivity.
  • Finger textured grip provides secure handling in wet or dry conditions
  • Ambidextrous design for quick and easy donning
  • Powder-free for clean comfort and superior tactile sensitivity
  • Beaded cuff enhances strength and prevents liquid run-off
  • Naturally anti-static properties (10^9 ohms)
  • Manufactured without plasticizers, silicone, or powders
  • RoHS compliant
  • Supplied in a cleanroom pack (100 gloves/pack)
Catalog No. 50-007-92250 Supplier MSC Supplier No. 96760475
These nitrile gloves are designed for use in controlled environments such as biotechnology and computer assembly, exceeding standards for Class 100 cleanrooms. They offer excellent grip and tactile sensitivity.
  • Finger textured grip provides secure handling in wet or dry conditions
  • Ambidextrous design for quick and easy donning
  • Powder-free for clean comfort and superior tactile sensitivity
  • Beaded cuff enhances strength and prevents liquid run-off
  • Naturally anti-static properties (10^9 ohms)
  • Manufactured without plasticizers, silicone, or powders
  • RoHS compliant
  • Supplied in a cleanroom pack (100 gloves/pack)