Learn More
MSC PIP Cold Condition Gloves, Wool-Lined, Large, Black, 1 Pair
Catalog No. 5000687955
Encompass
These Cold Condition Gloves feature a wool lining and knit wrist, providing thermal insulation and comfort for various cold environment applications.
  • Seamless construction for a comfortable fit.
  • Merino wool provides superior insulating properties even when wet and withstands moderately high temperatures.
  • Ambidextrous pattern permits the glove to be worn on either hand, extending wear and reducing costs.
  • Knit wrist helps prevent dirt and debris from entering the glove.
  • Excellent thermal insulation.
  • Quickly evaporates moisture from the skin.
Catalog No. 50-006-87955 Supplier MSC Supplier No. 08348146
These Cold Condition Gloves feature a wool lining and knit wrist, providing thermal insulation and comfort for various cold environment applications.
  • Seamless construction for a comfortable fit.
  • Merino wool provides superior insulating properties even when wet and withstands moderately high temperatures.
  • Ambidextrous pattern permits the glove to be worn on either hand, extending wear and reducing costs.
  • Knit wrist helps prevent dirt and debris from entering the glove.
  • Excellent thermal insulation.
  • Quickly evaporates moisture from the skin.