Learn More
Echelon Biosciences Research Labs PIP CONTROL BEADS 1 mL
SDP
Catalog No. 50720674
Encompass
Control Beads are blocked agarose beads for use as a negative control with Lipid Coated Beads. Echelons suite of Lipid Coated Beads are designed for protein pulldown experiments to identify novel protein-lipid interactions. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-674 Supplier Echelon Biosciences Research Labs Supplier No. PB0001ML
Control Beads are blocked agarose beads for use as a negative control with Lipid Coated Beads. Echelons suite of Lipid Coated Beads are designed for protein pulldown experiments to identify novel protein-lipid interactions. This item is non returnable