Learn More
MSC PIP Cut, Puncture & Abrasion-Resistant Gloves, A6 Cut, 18 Gauge, 2X-Large
Catalog No. 5000508743
Encompass
These cut, puncture, and abrasion-resistant gloves offer superior protection and dexterity. They feature an 18 gauge seamless knit lining and a polyurethane coating on the palm and fingertips for enhanced grip. Constructed from a blend of tungsten, carbon, elastane, HPPE, polyester, and polyurethane, these gloves are designed for high-performance applications.
  • High grip performance
  • Lightweight 18 gauge liner
  • OEKO-TEX 100 certified
  • Washable (3 washing cycles)
  • Low DMF content (less than 100 ppm)
  • Glass fiber free
  • Silicone free
  • Touchscreen capable
  • Antistatic
  • Certified for food handling
  • Breathable coating
  • FDA approved
Catalog No. 50-005-08743 Supplier MSC Supplier No. 41905589
These cut, puncture, and abrasion-resistant gloves offer superior protection and dexterity. They feature an 18 gauge seamless knit lining and a polyurethane coating on the palm and fingertips for enhanced grip. Constructed from a blend of tungsten, carbon, elastane, HPPE, polyester, and polyurethane, these gloves are designed for high-performance applications.
  • High grip performance
  • Lightweight 18 gauge liner
  • OEKO-TEX 100 certified
  • Washable (3 washing cycles)
  • Low DMF content (less than 100 ppm)
  • Glass fiber free
  • Silicone free
  • Touchscreen capable
  • Antistatic
  • Certified for food handling
  • Breathable coating
  • FDA approved