Learn More
MSC PIP Cut and Puncture-Resistant Sleeve, Nylon, Small, Black, 1 ea
Catalog No. 5000766685
Encompass
This cut and puncture-resistant sleeve provides protection for various industrial applications. Made from durable nylon, it features a hook and loop closure for a secure and comfortable fit, offering both cut resistance and breathability.
  • Hook and loop closure
  • Cut-resistant
  • Lightweight design
  • Medium duty protection
  • Smooth and soft texture
  • Breathable material
  • ANSI/ISEA Cut Resistance Level A4
  • Overall length 7.00 in.
Catalog No. 50-007-66685 Supplier MSC Supplier No. 50658178
This cut and puncture-resistant sleeve provides protection for various industrial applications. Made from durable nylon, it features a hook and loop closure for a secure and comfortable fit, offering both cut resistance and breathability.
  • Hook and loop closure
  • Cut-resistant
  • Lightweight design
  • Medium duty protection
  • Smooth and soft texture
  • Breathable material
  • ANSI/ISEA Cut Resistance Level A4
  • Overall length 7.00 in.