Learn More
MSC PIP Cut-Resistant Glove, ANSI A7, 18 Gauge, Foam Nitrile, Touchscreen
Catalog No. 5000508573
Encompass
This CoreShield™ cut-resistant glove offers optimal comfort, dexterity, and cut protection with its lightweight 18-gauge fiberglass-free liner. The foam nitrile coating on the palm and fingers provides enhanced grip in dry and slightly oily conditions. It is touchscreen compatible, ESD safe, and meets FDA food handling requirements.
  • Vend pack ready
  • Seamless construction for comfort and breathability
  • 18-gauge fiberglass-free liner for dexterity and cut resistance
  • Foam nitrile coating for improved grip
  • Thumb and forefinger reinforcement for durability
  • Touchscreen compatible
  • ESD safe
  • FDA food handling compliant (21 CFR 177.2600)
  • Silicone-free
  • Oeko-Tex 100 certified
  • Knit wrist design
Catalog No. 50-005-08573 Supplier MSC Supplier No. 37112976
This CoreShield™ cut-resistant glove offers optimal comfort, dexterity, and cut protection with its lightweight 18-gauge fiberglass-free liner. The foam nitrile coating on the palm and fingers provides enhanced grip in dry and slightly oily conditions. It is touchscreen compatible, ESD safe, and meets FDA food handling requirements.
  • Vend pack ready
  • Seamless construction for comfort and breathability
  • 18-gauge fiberglass-free liner for dexterity and cut resistance
  • Foam nitrile coating for improved grip
  • Thumb and forefinger reinforcement for durability
  • Touchscreen compatible
  • ESD safe
  • FDA food handling compliant (21 CFR 177.2600)
  • Silicone-free
  • Oeko-Tex 100 certified
  • Knit wrist design