This cut-resistant glove features a seamless knit construction for comfort, fit, and dexterity. Its Dyneema blend enhances performance and ensures maximum protection, while the ambidextrous pattern extends wear life and reduces costs.
- Seamless knit construction for comfort, fit, and dexterity
- Dyneema blend for enhanced performance and maximum protection
- Ambidextrous design for extended wear and reduced costs
- Special non-latex cuff reduces blowout even after multiple launderings
- Can be worn alone or as a glove liner
- Good gripping properties and shrink resistant
- Complies with FDA Food Handling Requirements 21 CFR, Part 177
- Antimicrobial, non-toxic, non-corrosive, and complies with FDA, EPA, and USDA Standards for Food Contact
- ANSI/ISEA Cut Resistance Level A7
- Primary material: Dyneema
- Size large (numeric size 9)
- FDA approved