Learn More
MSC PIP Cut Resistant Gloves, Cut A4, Puncture 2, 1 Pair
Catalog No. 5000803250
Encompass
These cut and puncture-resistant gloves offer robust hand protection, featuring durable cowhide leather construction and a para-aramid liner. Designed for applications requiring high safety against mechanical risks, they provide comfort and dexterity alongside essential protection.
  • Leather style with natural color and textured grip
  • ANSI/ISEA cut resistance level A4
  • ANSI/ISEA puncture resistance level 2
  • ANSI/ISEA abrasion resistance level 0
  • EN 388 rating 3X32DX
  • Para-aramid liner provides high tensile strength and inherent cut/heat resistance
  • Stitched with Kevlar for added strength and burn resistance
  • Keystone thumb for ergonomic fit and dexterity
  • Slip-on cuff style for good fit and comfort
  • ARC rating: ATPV 43 cal/cm² (PPE 4)
Catalog No. 50-008-03250 Supplier MSC Supplier No. 10657377
These cut and puncture-resistant gloves offer robust hand protection, featuring durable cowhide leather construction and a para-aramid liner. Designed for applications requiring high safety against mechanical risks, they provide comfort and dexterity alongside essential protection.
  • Leather style with natural color and textured grip
  • ANSI/ISEA cut resistance level A4
  • ANSI/ISEA puncture resistance level 2
  • ANSI/ISEA abrasion resistance level 0
  • EN 388 rating 3X32DX
  • Para-aramid liner provides high tensile strength and inherent cut/heat resistance
  • Stitched with Kevlar for added strength and burn resistance
  • Keystone thumb for ergonomic fit and dexterity
  • Slip-on cuff style for good fit and comfort
  • ARC rating: ATPV 43 cal/cm² (PPE 4)