Learn More
MSC PIP Cut-Resistant Gloves, PolyKor Nitrile, Hi-Vis Yellow, A3, 1 Pair
Catalog No. 5000766786
Encompass
G-Tek Barracuda Hi-Vis gloves feature a seamless knit PolyKor® blended shell for comfort, breathability, and cut resistance. The foam nitrile coating provides an improved grip by dispersing fluids on contact.
  • Seamless construction for increased comfort and breathability
  • Lightweight PolyKor® blended shell offers excellent dexterity, tactile sensitivity, and cut resistance
  • Breathable foam nitrile coating disperses fluids for improved grip
  • Silicone free
  • Knit wrist prevents dirt and debris entry
  • Hi-vis coloring for easy visibility
  • Secure dry grip and oil resistant
  • Ideal for handling/assembly of parts, metal fabrication, and equipment maintenance
Catalog No. 50-007-66786 Supplier MSC Supplier No. 63310486
G-Tek Barracuda Hi-Vis gloves feature a seamless knit PolyKor® blended shell for comfort, breathability, and cut resistance. The foam nitrile coating provides an improved grip by dispersing fluids on contact.
  • Seamless construction for increased comfort and breathability
  • Lightweight PolyKor® blended shell offers excellent dexterity, tactile sensitivity, and cut resistance
  • Breathable foam nitrile coating disperses fluids for improved grip
  • Silicone free
  • Knit wrist prevents dirt and debris entry
  • Hi-vis coloring for easy visibility
  • Secure dry grip and oil resistant
  • Ideal for handling/assembly of parts, metal fabrication, and equipment maintenance