These Barracuda Cut Resistant Gloves are designed for demanding work environments. Featuring HPPE fiber and a polyurethane palm, they provide superior cut protection and tactile grip for precise handling, resisting abrasion and cuts.
- Rugged HPPE fiber is 10 times stronger than steel and 40% stronger than blended fabrics
- Excellent cut protection in a comfortable glove
- A 13 ga gray HPPE fiber shell with gray polyurethane coating
- Seamless construction offers increased comfort and breathability
- Polyurethane coating provides a tactile grip in dry and slightly oily conditions, ideal for applications requiring dexterity
- Knit wrist helps prevent dirt and debris from entering the glove
- Color coded hems for easy size identification
- Washable, resistant to chemicals, water, and ultraviolet light