Learn More
MSC PIP Disposable Gloves, Ambi-Dex, Vinyl, Medium, 4 mil, 100/Pk.
Catalog No. 5000793700
Encompass
These disposable gloves are constructed from vinyl with a synthetic polymer coating, offering reliable hand protection for general purpose use. They feature an ambidextrous design, are non-sterile, and powdered for easy donning, making them suitable for a wide range of laboratory and industrial tasks.
  • Clear color
  • Length: 9.5 in
  • Smooth grip surface
  • Ambidextrous design
  • FDA approved for food contact
  • Non-sterile
  • Synthetic polymer coating
  • Nominal palm thickness: 4 mil
Catalog No. 50-007-93700 Supplier MSC Supplier No. 66897976
These disposable gloves are constructed from vinyl with a synthetic polymer coating, offering reliable hand protection for general purpose use. They feature an ambidextrous design, are non-sterile, and powdered for easy donning, making them suitable for a wide range of laboratory and industrial tasks.
  • Clear color
  • Length: 9.5 in
  • Smooth grip surface
  • Ambidextrous design
  • FDA approved for food contact
  • Non-sterile
  • Synthetic polymer coating
  • Nominal palm thickness: 4 mil