Learn More
MSC PIP Disposable Gloves, Nitrile, Medium, 8 mil, Powder-Free, 50 Pack
Catalog No. 5000802127
Encompass
These disposable nitrile gloves are designed for general purpose use, providing protection and dexterity for various tasks. They are powder-free and ambidextrous, ensuring comfort and ease of use in laboratory and industrial environments.
  • Nitrile material
  • Features 8 mil nominal palm thickness
  • General purpose grade
  • Has a 9.5 inch length
  • Medium size
  • Blue color
  • Non-sterile
  • Powder-free
  • Ambidextrous design
  • FDA approved
  • Textured finish
  • Packaged as 50 gloves per pack
Catalog No. 50-008-02127 Supplier MSC Supplier No. 72121668
These disposable nitrile gloves are designed for general purpose use, providing protection and dexterity for various tasks. They are powder-free and ambidextrous, ensuring comfort and ease of use in laboratory and industrial environments.
  • Nitrile material
  • Features 8 mil nominal palm thickness
  • General purpose grade
  • Has a 9.5 inch length
  • Medium size
  • Blue color
  • Non-sterile
  • Powder-free
  • Ambidextrous design
  • FDA approved
  • Textured finish
  • Packaged as 50 gloves per pack