Learn More
MSC PIP Drivers Gloves, Cowhide/Para-Aramid, ANSI Cut A4, 2X-Large, 1 Pair
Catalog No. 5000787160
Encompass
These drivers gloves provide protection from arc flash and feature slip-on styling for comfort. Constructed with top grain cowhide leather and a Para-Aramid liner, they offer excellent abrasion, cut, and heat resistance.
  • Slip-on styling for good fit and comfort
  • Top grain cowhide leather construction for durability and breathability
  • Para-Aramid liner provides high tensile strength and is inherently cut and heat resistant
  • Stitched with Kevlar for added strength and burn resistance
  • Keystone thumb design reduces hand fatigue and offers superior dexterity
  • Elastic closure sewn inside glove for safety
  • Arc rating: ATPV 43 cal/cm² PPE 4 (HRC 4)
  • ANSI cut level A4
  • ANSI puncture level 3
Catalog No. 50-007-87160 Supplier MSC Supplier No. 13629324
These drivers gloves provide protection from arc flash and feature slip-on styling for comfort. Constructed with top grain cowhide leather and a Para-Aramid liner, they offer excellent abrasion, cut, and heat resistance.
  • Slip-on styling for good fit and comfort
  • Top grain cowhide leather construction for durability and breathability
  • Para-Aramid liner provides high tensile strength and is inherently cut and heat resistant
  • Stitched with Kevlar for added strength and burn resistance
  • Keystone thumb design reduces hand fatigue and offers superior dexterity
  • Elastic closure sewn inside glove for safety
  • Arc rating: ATPV 43 cal/cm² PPE 4 (HRC 4)
  • ANSI cut level A4
  • ANSI puncture level 3