Learn More
MSC PIP Duster Raincoat, 60 in, Limited Flammability, 1 ea
Catalog No. 5000769438
Encompass
This 60-inch duster raincoat provides reliable protection against rain and offers limited flammability.
  • Treated to prevent after-flame
  • Completely waterproof
  • Features snap closure
  • Includes two pockets
  • Made from 0.35 mm PVC / Polyester FR treated fabric
  • Complies with ASTM D6413 for limited flammability
Catalog No. 50-007-69438 Supplier MSC Supplier No. 50307354
This 60-inch duster raincoat provides reliable protection against rain and offers limited flammability.
  • Treated to prevent after-flame
  • Completely waterproof
  • Features snap closure
  • Includes two pockets
  • Made from 0.35 mm PVC / Polyester FR treated fabric
  • Complies with ASTM D6413 for limited flammability