Learn More
MSC PIP Ear Plugs, 32 dB NRR, T-Shape, Uncorded, 200 Pairs/Pack
Catalog No. 5000792986
Encompass
These disposable earplugs are constructed from soft polyurethane foam and are designed to provide effective hearing protection. Featuring a T-shape and an uncorded design, they offer a Noise Reduction Rating (NRR) of 32 dB, suitable for various loud environments.
  • Innovative T-shape for simple removal
  • Consistent cellular structure for optimal expansion and a uniform fit
  • Smooth closed cell outer layer prevents debris build-up
  • PowerSoft™ foam provides soft, all-day comfort
  • Offers NRR 32 protection for effective noise reduction
Catalog No. 50-007-92986 Supplier MSC Supplier No. 98938269
These disposable earplugs are constructed from soft polyurethane foam and are designed to provide effective hearing protection. Featuring a T-shape and an uncorded design, they offer a Noise Reduction Rating (NRR) of 32 dB, suitable for various loud environments.
  • Innovative T-shape for simple removal
  • Consistent cellular structure for optimal expansion and a uniform fit
  • Smooth closed cell outer layer prevents debris build-up
  • PowerSoft™ foam provides soft, all-day comfort
  • Offers NRR 32 protection for effective noise reduction