Learn More
MSC PIP Earplugs, 30 dB NRR, Polyurethane, Corded, 100 Pairs/Pk.
Catalog No. 5000676366
Encompass
These earplugs provide effective hearing protection with a 30 dB noise reduction rating. Constructed from polyurethane with a cylinder shape, they feature a corded, push-in design for easy use. These disposable earplugs are also non-metal detectable.
  • Disposable design
  • Corded attachment
  • Cylinder plug shape
  • Noise reduction rating of 30 dB
  • Orange color
  • Polyurethane material
  • Push-in insertion
  • Universal size
  • Non-metal detectable
  • Black PVC cord
  • EZ-Twist series
  • Meets ANSI S3.19-1974 standards
Catalog No. 50-006-76366 Supplier MSC Supplier No. 14324685
These earplugs provide effective hearing protection with a 30 dB noise reduction rating. Constructed from polyurethane with a cylinder shape, they feature a corded, push-in design for easy use. These disposable earplugs are also non-metal detectable.
  • Disposable design
  • Corded attachment
  • Cylinder plug shape
  • Noise reduction rating of 30 dB
  • Orange color
  • Polyurethane material
  • Push-in insertion
  • Universal size
  • Non-metal detectable
  • Black PVC cord
  • EZ-Twist series
  • Meets ANSI S3.19-1974 standards