Learn More
MSC PIP Earplugs, 32 dB NRR, Polyurethane, Tapered Uncorded, 200 Pairs
Catalog No. 5000688552
Encompass
These disposable polyurethane earplugs provide a high noise reduction rating, featuring a tapered, roll-down design for easy insertion and a comfortable fit.
  • High noise reduction rating of 32 dB
  • Made from polyurethane for comfort and effective sealing
  • Tapered, roll-down design facilitates easy and secure insertion
  • Uncorded for freedom of movement and versatility
  • Universal size fits most users
  • Disposable for hygiene and convenience
  • Non-metal detectable, suitable for various environments
  • Meets ANSI S3.19-1974 standards
Catalog No. 50-006-88552 Supplier MSC Supplier No. 98938368
These disposable polyurethane earplugs provide a high noise reduction rating, featuring a tapered, roll-down design for easy insertion and a comfortable fit.
  • High noise reduction rating of 32 dB
  • Made from polyurethane for comfort and effective sealing
  • Tapered, roll-down design facilitates easy and secure insertion
  • Uncorded for freedom of movement and versatility
  • Universal size fits most users
  • Disposable for hygiene and convenience
  • Non-metal detectable, suitable for various environments
  • Meets ANSI S3.19-1974 standards