Learn More
Takara Bio PIP EIA KIT 96 RXNS
SDP
Supplier:  Takara Bio MK101
Encompass
Enzyme Immunoassay Kit; Procollagen Type I C-Peptide (PIP); Assay Range: 10-640ng/mL; Sensitivity: 10ng/mL; Sample Size: 20uL; 1 x 96 tests
Catalog No. 50-443-932