Learn More
MSC PIP Finger Cots, Anti-Static, Non-Powdered, Latex, X-Large, 720/Pack
Catalog No. 5000676177
Encompass
These anti-static latex finger cots are designed for use in EDS-sensitive areas and avionic applications, providing protection against electrostatic discharge.
  • Designed for EDS-sensitive area and avionic applications
  • Non-powdered
  • Made from latex
  • X-large size
  • Available in a 720 count pack
Catalog No. 50-006-76177 Supplier MSC Supplier No. 16040529
These anti-static latex finger cots are designed for use in EDS-sensitive areas and avionic applications, providing protection against electrostatic discharge.
  • Designed for EDS-sensitive area and avionic applications
  • Non-powdered
  • Made from latex
  • X-large size
  • Available in a 720 count pack