Learn More
MSC PIP G-Tek Perfect Fit Glove, A2 Cut, 18 Gauge, Polyurethane Coated
Catalog No. 5000508559
Encompass
These G-Tek Perfect Fit seamless knit gloves feature an 18-gauge engineered blend liner for dexterity, tactile sensitivity, and A2 cut resistance. The durable polyurethane coating provides superior tactility, abrasion resistance, and dry grip. They are touchscreen compatible and ESD safe.
  • Seamless construction offers increased comfort and breathability.
  • Lightweight 18-gauge engineered blend liner provides excellent dexterity, tactile sensitivity, and cut resistance.
  • Polyurethane is thin, yet durable and provides excellent tactility, abrasion resistance and grip on dry surfaces.
  • Touchscreen compatible to allow the user to operate a touch screen phone or device without removing gloves.
  • ESD safe construction makes these gloves safe for use with static sensitive materials.
  • Silicone free for smudge free handling.
Catalog No. 50-005-08559 Supplier MSC Supplier No. 05765052
These G-Tek Perfect Fit seamless knit gloves feature an 18-gauge engineered blend liner for dexterity, tactile sensitivity, and A2 cut resistance. The durable polyurethane coating provides superior tactility, abrasion resistance, and dry grip. They are touchscreen compatible and ESD safe.
  • Seamless construction offers increased comfort and breathability.
  • Lightweight 18-gauge engineered blend liner provides excellent dexterity, tactile sensitivity, and cut resistance.
  • Polyurethane is thin, yet durable and provides excellent tactility, abrasion resistance and grip on dry surfaces.
  • Touchscreen compatible to allow the user to operate a touch screen phone or device without removing gloves.
  • ESD safe construction makes these gloves safe for use with static sensitive materials.
  • Silicone free for smudge free handling.