Learn More
MSC PIP General Purpose Gloves, Cowhide Leather, Large, Natural, 1 Pair
Catalog No. 5000795669
Encompass
These general purpose gloves are crafted from cowhide leather with a cowhide leather lining, offering excellent abrasion resistance and general protection. They feature a slip-on design with an elastic closure and a straight thumb for comfort and a secure fit. Suitable for various applications including construction, utility work, and farming.
  • Top grain leather construction provides comfort, durability, and dexterity
  • Breathability
  • Slip-on styling for a good fit and comfort
  • Straight thumb for increased comfort in closed-fisted work
  • Elastic closure sewn inside glove for safety and comfort
  • Color coded hems for easy size identification
  • Excellent abrasion resistance
  • Full-fingered design
  • Nylon stitching
Catalog No. 50-007-95669 Supplier MSC Supplier No. 41720616
These general purpose gloves are crafted from cowhide leather with a cowhide leather lining, offering excellent abrasion resistance and general protection. They feature a slip-on design with an elastic closure and a straight thumb for comfort and a secure fit. Suitable for various applications including construction, utility work, and farming.
  • Top grain leather construction provides comfort, durability, and dexterity
  • Breathability
  • Slip-on styling for a good fit and comfort
  • Straight thumb for increased comfort in closed-fisted work
  • Elastic closure sewn inside glove for safety and comfort
  • Color coded hems for easy size identification
  • Excellent abrasion resistance
  • Full-fingered design
  • Nylon stitching