Learn More
MSC PIP General Purpose Gloves, Polyester, Polyester-Lined, Medium, White, 1 Pair
Catalog No. 5000767462
Encompass
These PIP general purpose gloves are made from polyester with a polyester lining, designed for various applications. They feature a seamless knit shell to reduce hand fatigue and offer abrasion resistance.
  • Seamless knit polyester shell reduces hand fatigue.
  • Low lint gloves knit from polyester filament yarns.
  • Can be worn under disposable and other liquid-proof gloves.
  • Provides abrasion resistance.
  • Polyester yarns can be washed with minimal shrinkage.
  • Polyester can be sterilized.
  • Silicone free.
Catalog No. 50-007-67462 Supplier MSC Supplier No. 18587485
These PIP general purpose gloves are made from polyester with a polyester lining, designed for various applications. They feature a seamless knit shell to reduce hand fatigue and offer abrasion resistance.
  • Seamless knit polyester shell reduces hand fatigue.
  • Low lint gloves knit from polyester filament yarns.
  • Can be worn under disposable and other liquid-proof gloves.
  • Provides abrasion resistance.
  • Polyester yarns can be washed with minimal shrinkage.
  • Polyester can be sterilized.
  • Silicone free.