Learn More
MSC PIP General Purpose Gloves, X-Large, Cotton, Open Cuff, 1 Pair
Catalog No. 5000793196
Encompass
These general purpose gloves are designed for comfort and light protection, featuring a cotton-lined, lint-free construction. They are suitable for various applications requiring general hand protection and comfort.
  • Cotton-lined and lint-free
  • Full fashion pattern with fourchettes and set-in thumbs for a tailored fit
  • Three lines of raised stitching on back with an open cuff finish
  • Seamless design
  • Dotted palm for improved grip
  • X-large size, full-fingered coverage
  • Unisex design
Catalog No. 50-007-93196 Supplier MSC Supplier No. 66864364
These general purpose gloves are designed for comfort and light protection, featuring a cotton-lined, lint-free construction. They are suitable for various applications requiring general hand protection and comfort.
  • Cotton-lined and lint-free
  • Full fashion pattern with fourchettes and set-in thumbs for a tailored fit
  • Three lines of raised stitching on back with an open cuff finish
  • Seamless design
  • Dotted palm for improved grip
  • X-large size, full-fingered coverage
  • Unisex design