Learn More
MSC PIP Global Cut Resistant Gloves, X-Small, HPPE, Polyurethane, A2
Catalog No. 5000510025
Encompass
These gloves provide lightweight protection and tactility for precise handling of small parts. They feature an 18-gauge liner and a thin, durable polyurethane coating, making them suitable for automotive, manufacturing, paper & pulp, aerospace, and food processing applications.
  • Seamless construction for comfort and breathability.
  • Lightweight 18-gauge engineered blend liner offers excellent dexterity, tactile sensitivity, and cut resistance.
  • Polyurethane coating provides tactility, abrasion resistance, and grip on dry surfaces.
  • Touchscreen compatible.
  • ESD safe construction for use with static-sensitive materials.
  • Silicone-free for smudge-free handling.
  • Complies with FDA food handling requirements.
  • Knit wrist helps prevent dirt and debris.
Catalog No. 50-005-10025 Supplier MSC Supplier No. 05764402
These gloves provide lightweight protection and tactility for precise handling of small parts. They feature an 18-gauge liner and a thin, durable polyurethane coating, making them suitable for automotive, manufacturing, paper & pulp, aerospace, and food processing applications.
  • Seamless construction for comfort and breathability.
  • Lightweight 18-gauge engineered blend liner offers excellent dexterity, tactile sensitivity, and cut resistance.
  • Polyurethane coating provides tactility, abrasion resistance, and grip on dry surfaces.
  • Touchscreen compatible.
  • ESD safe construction for use with static-sensitive materials.
  • Silicone-free for smudge-free handling.
  • Complies with FDA food handling requirements.
  • Knit wrist helps prevent dirt and debris.