Learn More
MSC PIP Glove, Kevlar, ANSI Cut A2, Universal Size, 1 ea
Catalog No. 5000787381
Encompass
This cut-resistant glove features a universal size and a continuous knit cuff, designed for protection in various applications.
  • Made from Kevlar material
  • Brown color
  • Continuous knit cuff closure
  • Overall length 26.00 in
  • Includes a thumb hole
  • Cut-resistant features
  • ANSI/ISEA abrasion resistance level 2
  • ANSI/ISEA cut resistance level A2
Catalog No. 50-007-87381 Supplier MSC Supplier No. 52576261
This cut-resistant glove features a universal size and a continuous knit cuff, designed for protection in various applications.
  • Made from Kevlar material
  • Brown color
  • Continuous knit cuff closure
  • Overall length 26.00 in
  • Includes a thumb hole
  • Cut-resistant features
  • ANSI/ISEA abrasion resistance level 2
  • ANSI/ISEA cut resistance level A2