Learn More
MSC PIP Gloves, Cut A4, Polyurethane-Coated Palm, 21 Gauge, 1 Pair
Catalog No. 5000796923
Encompass
These G-Tek® 16-541 gloves offer cut, puncture, and abrasion resistance, featuring a PolyKor, polyurethane-coated palm and fingers for enhanced grip and protection. Their seamless, lightweight design ensures comfort, dexterity, and tactile sensitivity for various demanding applications.
  • Seamless construction offers increased comfort and breathability
  • Premium PolyKor blended 21 gauge shell is lightweight
  • Provides excellent dexterity and tactile sensitivity
  • Features a polyurethane-coated palm and fingers
  • ANSI/ISEA A4 cut resistance level
  • ANSI/ISEA 2 puncture resistance level
  • ANSI/ISEA 3 abrasion resistance level
Catalog No. 50-007-96923 Supplier MSC Supplier No. 13629142
These G-Tek® 16-541 gloves offer cut, puncture, and abrasion resistance, featuring a PolyKor, polyurethane-coated palm and fingers for enhanced grip and protection. Their seamless, lightweight design ensures comfort, dexterity, and tactile sensitivity for various demanding applications.
  • Seamless construction offers increased comfort and breathability
  • Premium PolyKor blended 21 gauge shell is lightweight
  • Provides excellent dexterity and tactile sensitivity
  • Features a polyurethane-coated palm and fingers
  • ANSI/ISEA A4 cut resistance level
  • ANSI/ISEA 2 puncture resistance level
  • ANSI/ISEA 3 abrasion resistance level