Learn More
MSC PIP Gloves, Cut A4, Puncture 3, Abrasion 6
Catalog No. 5000509590
Encompass
These cut, puncture, and abrasion-resistant gloves are engineered to be lightweight and glass fiber-free. They feature a specialty engineered polyurethane (PU) coating designed with a low level of Dimethylformamide (DMF) to enhance worker protection.
  • Cut resistance level A4 (ANSI/ISEA).
  • Puncture resistance level 3 (ANSI/ISEA).
  • Abrasion resistance level 6 (ANSI/ISEA).
  • 13 gauge construction.
  • Primary material: HPPE.
  • Polyurethane coating on palm and fingertips.
  • Smooth grip surface.
  • Knit cuff style.
  • EN 388 rating: 4X44D.
  • Meets standards: EU 1935/2004, ANSI 105, ENISO 21420, EN388, ANSI 105:2016.
Catalog No. 50-005-09590 Supplier MSC Supplier No. 97297659
These cut, puncture, and abrasion-resistant gloves are engineered to be lightweight and glass fiber-free. They feature a specialty engineered polyurethane (PU) coating designed with a low level of Dimethylformamide (DMF) to enhance worker protection.
  • Cut resistance level A4 (ANSI/ISEA).
  • Puncture resistance level 3 (ANSI/ISEA).
  • Abrasion resistance level 6 (ANSI/ISEA).
  • 13 gauge construction.
  • Primary material: HPPE.
  • Polyurethane coating on palm and fingertips.
  • Smooth grip surface.
  • Knit cuff style.
  • EN 388 rating: 4X44D.
  • Meets standards: EU 1935/2004, ANSI 105, ENISO 21420, EN388, ANSI 105:2016.