Learn More
MSC PIP Gloves, Dyneema, Cut A2, 13 ga, Small
Catalog No. 5000509267
Encompass
These cut, puncture, and abrasion-resistant gloves are engineered to be lightweight and glass fiber-free, offering superior protection. They feature a specially engineered polyurethane coating on the palm and fingertips, designed with a low level of dimethylformamide (DMF) to enhance worker safety. Made with Dyneema material, these gloves provide excellent durability and a smooth grip.
  • Lightweight and glass fiber-free construction
  • Polyurethane coating on palm and fingertips
  • Low dimethylformamide (DMF) level in coating
  • Dyneema primary material for cut, puncture, and abrasion resistance
  • ANSI/ISEA cut resistance level A2
  • ANSI/ISEA abrasion resistance level 6
  • ANSI/ISEA puncture resistance level 2
  • 13 gauge for dexterity
  • Smooth grip surface
  • Seamless knit style
  • White color
  • Unisex design
Catalog No. 50-005-09267 Supplier MSC Supplier No. 97380943
These cut, puncture, and abrasion-resistant gloves are engineered to be lightweight and glass fiber-free, offering superior protection. They feature a specially engineered polyurethane coating on the palm and fingertips, designed with a low level of dimethylformamide (DMF) to enhance worker safety. Made with Dyneema material, these gloves provide excellent durability and a smooth grip.
  • Lightweight and glass fiber-free construction
  • Polyurethane coating on palm and fingertips
  • Low dimethylformamide (DMF) level in coating
  • Dyneema primary material for cut, puncture, and abrasion resistance
  • ANSI/ISEA cut resistance level A2
  • ANSI/ISEA abrasion resistance level 6
  • ANSI/ISEA puncture resistance level 2
  • 13 gauge for dexterity
  • Smooth grip surface
  • Seamless knit style
  • White color
  • Unisex design