Learn More
MSC PIP Gloves, Nitrile, 15 mil, Raised Diamond Grip, 1 Pair
Catalog No. 5000803518
Encompass
Assurance chemical-resistant gloves are constructed from nitrile for reliable protection. Designed for various applications including chemical processing and food handling, these unlined gloves feature a raised diamond grip and straight cuff, meeting FDA and EN standards.
  • Offers resistance to abrasion, cuts, and punctures
  • Medium-weight construction for comfort and protection
  • Provides flexibility, tactile sensitivity, and dexterity
  • Protects from chemical and physical hazards
  • Unlined to reduce product contamination
  • Straight cuff protects forearms from liquid run-off
  • Raised diamond pattern for enhanced grip
  • Chlorinated to harden and cleanse the surface
  • Complies with FDA food handling requirements
  • Silicone free
Catalog No. 50-008-03518 Supplier MSC Supplier No. 48015861
Assurance chemical-resistant gloves are constructed from nitrile for reliable protection. Designed for various applications including chemical processing and food handling, these unlined gloves feature a raised diamond grip and straight cuff, meeting FDA and EN standards.
  • Offers resistance to abrasion, cuts, and punctures
  • Medium-weight construction for comfort and protection
  • Provides flexibility, tactile sensitivity, and dexterity
  • Protects from chemical and physical hazards
  • Unlined to reduce product contamination
  • Straight cuff protects forearms from liquid run-off
  • Raised diamond pattern for enhanced grip
  • Chlorinated to harden and cleanse the surface
  • Complies with FDA food handling requirements
  • Silicone free