Learn More
MSC PIP Gloves, Nitrile Coated Nylon, ANSI/ISEA Cut A1, X-Small
Catalog No. 5000510035
Encompass
These gloves are designed to provide cut, puncture, and abrasion resistance. They are made of nylon with a nitrile coating on the palm and fingertips, featuring a smooth grip.
  • Enhanced cut resistance
  • Comfort
  • Ease of selection
  • Cost effectiveness
  • Touchscreen palm
  • Reinforced coating
Catalog No. 50-005-10035 Supplier MSC Supplier No. 10321552
These gloves are designed to provide cut, puncture, and abrasion resistance. They are made of nylon with a nitrile coating on the palm and fingertips, featuring a smooth grip.
  • Enhanced cut resistance
  • Comfort
  • Ease of selection
  • Cost effectiveness
  • Touchscreen palm
  • Reinforced coating