Learn More
MSC PIP HardCap A1 Bump Cap, Short Brim, Black, 1 ea
Catalog No. 5000767450
Encompass
The PIP HardCap A1+ Baseball Style Bump Cap features an ultra-sleek design that is nearly indistinguishable from a standard low-profile baseball cap. It incorporates a removable High-Density Polyethylene (HDPE) protective liner, which enables the outer cap to be fully washable. The cap also includes a unique size adjuster with an elastic strap for a quick and secure fit, angled to comfortably rest under the nape of the neck.
  • Removable protective HDPE liner for easy washing
  • Unique size adjuster on an elastic strip for a quick, secure fit
  • Terry cloth sweatband for comfort and absorption
  • Reflective piping for extra visibility and safety
  • Liner holes and mesh panels for added ventilation
Catalog No. 50-007-67450 Supplier MSC Supplier No. 68127943
The PIP HardCap A1+ Baseball Style Bump Cap features an ultra-sleek design that is nearly indistinguishable from a standard low-profile baseball cap. It incorporates a removable High-Density Polyethylene (HDPE) protective liner, which enables the outer cap to be fully washable. The cap also includes a unique size adjuster with an elastic strap for a quick and secure fit, angled to comfortably rest under the nape of the neck.
  • Removable protective HDPE liner for easy washing
  • Unique size adjuster on an elastic strip for a quick, secure fit
  • Terry cloth sweatband for comfort and absorption
  • Reflective piping for extra visibility and safety
  • Liner holes and mesh panels for added ventilation