Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC PIP High-Visibility T-Shirt, Medium, Polyester, Class 2, Black/Yellow, 1 ea

Catalog No. 5000794289
Encompass

This high-visibility work shirt is a T-shirt style garment made from polyester material. It is designed for enhanced safety and comfort in various work environments.

  • Made from polyester material
  • T-shirt style garment
  • Features zipper closure
  • Provides 50+ UPF sun protection
  • Class 2 hazardous protection level
  • Color: black and high-visibility yellow
  • Size: medium

Catalog No. 50-007-94289 Supplier MSC Supplier No. 48064026
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This high-visibility work shirt is a T-shirt style garment made from polyester material. It is designed for enhanced safety and comfort in various work environments.

  • Made from polyester material
  • T-shirt style garment
  • Features zipper closure
  • Provides 50+ UPF sun protection
  • Class 2 hazardous protection level
  • Color: black and high-visibility yellow
  • Size: medium

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.