Learn More
MSC PIP High Visibility Vest, 2X-Large, ANSI Class 2, 1 ea
Catalog No. 5000794504
Encompass
This high visibility vest is designed for general purpose use, offering enhanced visibility in various work environments. It features a lightweight mesh construction and is classified as ANSI Class 2 for safety compliance.
  • Size: 2X-Large
  • Color: Orange
  • Closure type: Hook & loop
  • Pockets: 2
  • Material: Mesh
  • Horizontal stripes: 1
  • Stripe color: Silver
  • Stripe width: 2 in
  • Unisex design
  • ANSI Class 2 compliant
Catalog No. 50-007-94504 Supplier MSC Supplier No. 48017826
This high visibility vest is designed for general purpose use, offering enhanced visibility in various work environments. It features a lightweight mesh construction and is classified as ANSI Class 2 for safety compliance.
  • Size: 2X-Large
  • Color: Orange
  • Closure type: Hook & loop
  • Pockets: 2
  • Material: Mesh
  • Horizontal stripes: 1
  • Stripe color: Silver
  • Stripe width: 2 in
  • Unisex design
  • ANSI Class 2 compliant