Learn More
MSC PIP High Visibility Vest, Large, ANSI 2, Orange, 1 ea
Catalog No. 5000803550
Encompass
This high visibility vest is designed for general purpose use, meeting ANSI Class 2 standards. It features a Hi-Vis Orange color, a zipper closure, and is constructed from breathable mesh material. The vest includes two pockets and a single 2 inch silver horizontal stripe, suitable for unisex wear.
  • Large size
  • Hi-vis orange color
  • Zipper closure
  • Two pockets
  • Breathable mesh material
  • One 2 inch silver horizontal stripe
  • ANSI Class 2 compliant
  • Unisex design
Catalog No. 50-008-03550 Supplier MSC Supplier No. 48021174
This high visibility vest is designed for general purpose use, meeting ANSI Class 2 standards. It features a Hi-Vis Orange color, a zipper closure, and is constructed from breathable mesh material. The vest includes two pockets and a single 2 inch silver horizontal stripe, suitable for unisex wear.
  • Large size
  • Hi-vis orange color
  • Zipper closure
  • Two pockets
  • Breathable mesh material
  • One 2 inch silver horizontal stripe
  • ANSI Class 2 compliant
  • Unisex design