Learn More
MSC PIP HPPE Nitrile Foam Gloves, ANSI Cut A3, 1 Pair
Catalog No. 5000675709
Encompass
These seamless knit HPPE blended gloves are designed for applications requiring cut resistance and dexterity, such as electronics, glass cutting, and parts assembly.
  • Seamless construction offers increased comfort and breathability.
  • HPPE blended shell provides excellent dexterity, tactile sensitivity, and cut resistance.
  • Foam nitrile coatings disperse fluids for improved grip and are breathable and durable.
  • Knit wrist helps prevent dirt and debris from entering the glove.
  • Color coded hems for easy size identification.
  • Washable, resistant to chemicals, water, and ultraviolet light.
  • Features a 15 gauge knit for fine work.
  • Touchscreen compatible.
  • Silicone free.
  • Latex free.
  • OEKO-TEX certified.
  • Dermatological accreditation.
  • ANSI abrasion level 3 (1,900 cycles).
Catalog No. 50-006-75709 Supplier MSC Supplier No. 83805481
These seamless knit HPPE blended gloves are designed for applications requiring cut resistance and dexterity, such as electronics, glass cutting, and parts assembly.
  • Seamless construction offers increased comfort and breathability.
  • HPPE blended shell provides excellent dexterity, tactile sensitivity, and cut resistance.
  • Foam nitrile coatings disperse fluids for improved grip and are breathable and durable.
  • Knit wrist helps prevent dirt and debris from entering the glove.
  • Color coded hems for easy size identification.
  • Washable, resistant to chemicals, water, and ultraviolet light.
  • Features a 15 gauge knit for fine work.
  • Touchscreen compatible.
  • Silicone free.
  • Latex free.
  • OEKO-TEX certified.
  • Dermatological accreditation.
  • ANSI abrasion level 3 (1,900 cycles).