Learn More
MSC PIP MaxiCut 3 Gloves, EN388 Cut 3, 1 Pair
Catalog No. 5000768464
Encompass
The MaxiCut 3 gloves feature an engineered yarn shell with a MicroFoam nitrile coated palm and finger tips. Designed for cut protection, these gloves are washable, chemical, water, and UV light resistant, and include a reinforced thumb crotch for enhanced durability.
  • Washable and resistant to chemicals, water, and ultraviolet light
  • Seamless construction
  • Nitrile coating provides good grip in oily and wet conditions
  • Reinforced thumb crotch
  • Contains no stainless steel fibers
  • Used for electronics, glass cutting, metal handling and fabrication, food processing, and meat processing
Catalog No. 50-007-68464 Supplier MSC Supplier No. 45641693
The MaxiCut 3 gloves feature an engineered yarn shell with a MicroFoam nitrile coated palm and finger tips. Designed for cut protection, these gloves are washable, chemical, water, and UV light resistant, and include a reinforced thumb crotch for enhanced durability.
  • Washable and resistant to chemicals, water, and ultraviolet light
  • Seamless construction
  • Nitrile coating provides good grip in oily and wet conditions
  • Reinforced thumb crotch
  • Contains no stainless steel fibers
  • Used for electronics, glass cutting, metal handling and fabrication, food processing, and meat processing