Learn More
MSC PIP North Half Mask Respirator, Small, Silicone
Catalog No. 5000507287
Encompass
The PIP North RU8800 Half Mask Respirator is designed for superior fit, comfort, and protection, particularly for hard-to-fit users. Featuring a soft and flexible silicone facepiece and a triple flange design with three distinct sealing surfaces, it ensures security and greater freedom of movement for all-day use.
  • Triple flange design provides superior fit, comfort, and protection
  • Accommodates a wide variety of face sizes, including extra small or petite individuals
  • Wraparound profile forms to the user's face for a better seal
  • Low placement of inhalation valves maximizes the field of vision
  • Exhalation valve provides quick exhaust of warm, exhaled air
  • Headstrap assembly includes an extra wide cradle suspension for even weight distribution
  • Woven straps are easy to adjust and secure
  • Niosh approved
Catalog No. 50-005-07287 Supplier MSC Supplier No. 82609447
The PIP North RU8800 Half Mask Respirator is designed for superior fit, comfort, and protection, particularly for hard-to-fit users. Featuring a soft and flexible silicone facepiece and a triple flange design with three distinct sealing surfaces, it ensures security and greater freedom of movement for all-day use.
  • Triple flange design provides superior fit, comfort, and protection
  • Accommodates a wide variety of face sizes, including extra small or petite individuals
  • Wraparound profile forms to the user's face for a better seal
  • Low placement of inhalation valves maximizes the field of vision
  • Exhalation valve provides quick exhaust of warm, exhaled air
  • Headstrap assembly includes an extra wide cradle suspension for even weight distribution
  • Woven straps are easy to adjust and secure
  • Niosh approved