This G-Tek PolyKor Seamless Knit A3 glove provides comfort and protection for industrial tasks. Its lightweight PolyKor blended shell offers excellent dexterity, tactile sensitivity, and cut resistance. The Nitrile MicroSurface coating ensures an elevated grip in both dry and oily conditions, with tiny suction cup pockets that disperse fluids on contact. A knit wrist prevents dirt and debris from entering, and color-coded hems allow for easy size identification.
- Seamless construction for increased comfort and breathability.
- Lightweight PolyKor blended shell provides excellent dexterity, tactile sensitivity, and cut resistance.
- Nitrile MicroSurface coating ensures superior grip in dry and oily conditions.
- Knit wrist design prevents entry of dirt and debris.
- Color-coded hems for quick size identification.
- Silicone-free construction.