Learn More
MSC PIP Protective Gloves, Cut A4, Puncture 2, 3X-Large, 1 ea
Catalog No. 5000770293
Encompass
These cut and puncture-resistant gloves offer robust protection with ANSI/ISEA Cut Resistance Level A4, Puncture Resistance Level 2, and Abrasion Resistance Level 0. Constructed from top-grain cowhide leather, they provide comfort, durability, and breathability. A para-aramid liner ensures high tensile strength, cut, and heat resistance, enhanced by Kevlar stitching and an ergonomic Keystone thumb design.
  • Arc rating: ATPV 43 cal/cm2, PPE 4 (HRC 4)
  • Slip-on styling provides a good fit and comfort
  • Top grain cowhide leather construction for comfort, durability, and abrasion resistance
  • Para-aramid liner for high tensile strength, cut, and heat resistance
  • Kevlar stitching adds strength and burn resistance
  • Keystone thumb design reduces hand fatigue, improves dexterity and durability
  • Elastic closure sewn inside for safety and comfort
Catalog No. 50-007-70293 Supplier MSC Supplier No. 15191844
These cut and puncture-resistant gloves offer robust protection with ANSI/ISEA Cut Resistance Level A4, Puncture Resistance Level 2, and Abrasion Resistance Level 0. Constructed from top-grain cowhide leather, they provide comfort, durability, and breathability. A para-aramid liner ensures high tensile strength, cut, and heat resistance, enhanced by Kevlar stitching and an ergonomic Keystone thumb design.
  • Arc rating: ATPV 43 cal/cm2, PPE 4 (HRC 4)
  • Slip-on styling provides a good fit and comfort
  • Top grain cowhide leather construction for comfort, durability, and abrasion resistance
  • Para-aramid liner for high tensile strength, cut, and heat resistance
  • Kevlar stitching adds strength and burn resistance
  • Keystone thumb design reduces hand fatigue, improves dexterity and durability
  • Elastic closure sewn inside for safety and comfort