Learn More
MSC PIP PVC Dipped Glove, Rough Finish, 10 in Length, 12/Pk.
Catalog No. 5000802957
Encompass
These PIP PVC dipped gloves are designed for demanding applications in industries such as mining, construction, refining, and petrochemicals. They provide protection when handling oily materials and are chemical-resistant, offering defense against abrasion, acids, grease, and solvents.
  • Economical enhanced wet grip and cut resistance with rough finish styles
  • Great dexterity, quick drying, and low lint shedding due to interlock lining
  • Black color helps hide dirt, maintaining appearance in harsh environments
  • Single dip construction for consistent protection
  • Straight cuff provides additional length for forearm protection against liquid run-off
  • Cotton knit interlock lining wicks away moisture for comfort and keeps hands dry
Catalog No. 50-008-02957 Supplier MSC Supplier No. 03594165
These PIP PVC dipped gloves are designed for demanding applications in industries such as mining, construction, refining, and petrochemicals. They provide protection when handling oily materials and are chemical-resistant, offering defense against abrasion, acids, grease, and solvents.
  • Economical enhanced wet grip and cut resistance with rough finish styles
  • Great dexterity, quick drying, and low lint shedding due to interlock lining
  • Black color helps hide dirt, maintaining appearance in harsh environments
  • Single dip construction for consistent protection
  • Straight cuff provides additional length for forearm protection against liquid run-off
  • Cotton knit interlock lining wicks away moisture for comfort and keeps hands dry