Learn More
MSC PIP Safety Glasses, Scratch-Resistant, Polycarbonate, Clear Lenses, Frameless, 1 ea
Catalog No. NC3965076
Encompass
These safety glasses feature a frameless design with scratch-resistant, clear polycarbonate lenses, providing essential eye protection. They are designed with a clear and pink frame and non-adjustable temples, part of the Eva Petite series, and offer integrated UV protection for various work environments.
  • Frameless design for a wide field of vision
  • Scratch-resistant lens coating for enhanced durability
  • Clear polycarbonate lenses ensure optical clarity
  • Integrated UV protection guards against harmful rays
  • Complies with ANSI Z87.1+ standards for safety
  • Small size for a comfortable fit
  • Wrap around frame design for comprehensive coverage
Catalog No. NC3965076 Supplier MSC Supplier No. 74589722
These safety glasses feature a frameless design with scratch-resistant, clear polycarbonate lenses, providing essential eye protection. They are designed with a clear and pink frame and non-adjustable temples, part of the Eva Petite series, and offer integrated UV protection for various work environments.
  • Frameless design for a wide field of vision
  • Scratch-resistant lens coating for enhanced durability
  • Clear polycarbonate lenses ensure optical clarity
  • Integrated UV protection guards against harmful rays
  • Complies with ANSI Z87.1+ standards for safety
  • Small size for a comfortable fit
  • Wrap around frame design for comprehensive coverage